盐酸胍 8 mol/l aqueous solution

供应商: Thermo Fisher Scientific
Warning

Synonyms: 盐酸胍, 亚氨基脲, 亚氨脲

24115
PIER24115EA 0 CNY
PIER24115
盐酸胍 8 mol/l aqueous solution
盐酸胍
分子式: NH₂C(=NH)NH₂·HCl
分子量: 95.53 g/mol
溶点: 185
存储条件: Ambient
MDL编号: MFCD00013026
CAS号: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR