SDS precipitation kit, SDS-Out™

供应商: Thermo Fisher Scientific

20308
PIER20308EA 0 CNY
PIER20308
SDS precipitation kit, SDS-Out™
蛋白质提取系统 蛋白提取试剂

SDS-Out™ SDS Precipitation Kit provides convenient removal of SDS from small-volume (0,1 to 10 ml) protein samples.


  • Convenient removal of SDS from microliter-volume samples
  • Used for samples containing 0,1 to 1% SDS
  • Minimal dilution of small-volume samples
  • Protein recovery is 90 to 100%


Delivery information: The kit includes SDS precipitation reagent (10 ml), spin cup columns (12 columns) and collection tubes (12 tubes).

现在订购

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR