Nitrogen generator, Solaris
供应商: PEAK SCIENTIFIC INSTRUMENTS
65-0001
PEAK65-0001EA 0 CNY
PEAK65-0001
Nitrogen generator, Solaris
气体发生器 氮发生器
This nitrogen generator has been engineered and designed as a gas delivery solution for ELSD, Compact Mass Spectrometer instruments and TLD readers which can reduce downtime and increase workflow efficiencies.
  • Space saving design
  • Variable outlet flow, pressure and purity
  • Optional Solaris air compressor stackable module
  • Colour indicative LED lighting for easy status awareness
The generator can be placed on a bench top and paired with an additional air compressor unit to provide air supply for labs without an in-house air supply or those who wish toMore Product Information