VWR® Carriers for Biological Specimens
供应商: VWR Collection
PBIB23144EA 0 CNY
PBIB23144
VWR® Carriers for Biological Specimens
搬运篮 瓶载体/容器
Sturdy container for transport of biological samples taken at patient’s home.
  • Foldaway handle for easy stackability of several units
  • Lugs to facilitate stacking and removable lid to facilitate cleaning
  • Biohazard symbol; personalisable with own data/logo
  • Shock-proof construction in lightweight material
  • Equipped with special seal to guarantee carrier remains airtight conforming with safety standards
  • Emergency procedures are printed on the side in the event of container breakage