Avantor SterilEnz®-II/AT: pre-gasketed sanitary fittings for single-use systems
供应商: PAW BIOSCIENCE
PAWB2AT-616-TC-10EA 0 CNY
PAWB2AT-616-TC-10 PAWB2AT-812-TC-10 PAWB2AT-816-TC-10 PAWB2AT-1216-TC-10 PAWB2AT-408-TC-10 PAWB2AT-416-TC-10 PAWB2AT-608-TC-10
Avantor SterilEnz®-II/AT: pre-gasketed sanitary fittings for single-use systems
接头
SterilEnz®-II/AT connectors come with a platinum cured, medical grade silicone gasket mechanically attached to the fitting face. They are designed for use in irradiated disposable systems.
  • Standard sanitary clamp fitting, per ASME BPE Specifications
  • Class VI Compliant materials; certificate included
  • Won't leak or blow-off like stepped barbs can
  • Mono-barb design
  • Zero mold parting lines on all critical sealing surfaces
  • Superior tooling
  • Gamma-stable, 25 to 40 kGY
Certifications: USP <88>: Class VI Biological reactivity tests, in vivo
USP <87>:More Product Information