Centrifugal devices, Nanosep® and Nanosep® MF
供应商: CYTIVA PALL
ODM02C34 OD003C33 OD003C34 OD003C35 OD010C33 OD010C34 OD010C35 OD030C33 OD030C34 OD030C35 OD100C33 OD100C34 OD100C35 OD300C33 OD300C34 OD300C35 ODM02C33 ODM02C35 ODM45C34 ODM45C35
PALLODM02C34EA 0 CNY
PALLODM02C34 PALLOD003C33 PALLOD003C34 PALLOD003C35 PALLOD010C33 PALLOD010C34 PALLOD010C35 PALLOD030C33 PALLOD030C34 PALLOD030C35 PALLOD100C33 PALLOD100C34 PALLOD100C35 PALLOD300C33 PALLOD300C34 PALLOD300C35 PALLODM02C33 PALLODM02C35 PALLODM45C34 PALLODM45C35
Centrifugal devices, Nanosep® and Nanosep® MF
过滤器 离心过滤器
These devices are used for simple, reliable concentrating and desalting of 50 to 500 μl samples. Select the low protein binding Omega™ membrane for ultrafiltration applications. The Bio-Inert® membrane is ideal for microfiltration of biological solutions. The GHP membrane is ideal for sample preparation in analytical chemistry (HPLC, GC, IC and MS). It is suitable for aqueous, organic and aggressive organic samples, and for rapid processing of samples.
  • Rapid processing of samples
  • Typical recoveries are greater than 90%
Housing: Low binding PP.