Tangential flow filtration capsules, Minimate™
供应商: CYTIVA PALL
OA010C12 OA030C12 OA100C12 OA001C12 OA003C12 OA050C12 OA070C12 OA300C12 OA500C12 OA990C12 OA005C12
PALLOA010C12EA 0 CNY
PALLOA010C12 PALLOA030C12 PALLOA100C12 PALLOA001C12 PALLOA003C12 PALLOA050C12 PALLOA070C12 PALLOA300C12 PALLOA500C12 PALLOA990C12 PALLOA005C12
Tangential flow filtration capsules, Minimate™
过滤器 切向流盒
The disposable Minimate™ TFF capsules, with Omega™ membrane, can perform concentration processes desalting or buffer exchange on the same system with minimal user intervention, and achieves high concentration factors in a single step due to the low hold-up volume of the devices.
  • Scaleable
  • Works with a variety of positive pressure laboratory pumps
  • High product recovery
  • 100% integrity tested
The capsules will process samples of up to 1 litre, or efficiently concentrate samples as little as 5 ml.
Delivery information: Supplied with all fittings andMore Product Information