Filter plates, 96-well, for solvent filtration, AcroPrep™ Advance
供应商: CYTIVA PALL
8047 8048 8247 8148 8147 8248
PALL8047EA 0 CNY
PALL8047 PALL8048 PALL8247 PALL8148 PALL8147 PALL8248
Filter plates, 96-well, for solvent filtration, AcroPrep™ Advance
微孔板 过滤板
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes that provide the best possible performance for a wide range of sample preparation and high throughput screening applications. Available with a choice of PTFE or PTFE membrane.
  • Chemically resistant materials provide a stable platform to process samples in organic solvents
  • Low, non specific binding: inert construction materials ensure complete sample recovery
  • Well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up
More Product Information