Filter plates, 96-well, for ultrafiltration, AcroPrep™ Advance
供应商: CYTIVA PALL
8034 8036 8035 8033 8166 8165 8164 8163
PALL8034EA 0 CNY
PALL8034 PALL8036 PALL8035 PALL8033 PALL8166 PALL8165 PALL8164 PALL8163
Filter plates, 96-well, for ultrafiltration, AcroPrep™ Advance
微孔板 过滤板
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides; bead and chromatography resin-based applications, gross fractionation and lysate clarification and general filtration.
  • High biomolecule recovery - Omega® membrane typically results in ≥90% recovery of target biomolecules
  • Well
More Product Information