Filter plates, 96-well, for multiplexing, AcroPrep™ Advance
供应商: CYTIVA PALL
8019 8049 8029 8027
PALL8019EA 0 CNY
PALL8019 PALL8049 PALL8029 PALL8027
Filter plates, 96-well, for multiplexing, AcroPrep™ Advance
微孔板 过滤板
The AcroPrep™ Advance filter plate portfolio includes a variety of membrane types and pore sizes to provide the best possible performance for a wide range of sample preparation and high throughput screening applications. AcroPrep™ Advance filter plates are ideal for concentration, purification and desalting of proteins and peptides, bead and chromatography resin-based applications, gross fractionation and lysate clarification, and general filtration.
  • Smooth well wall provides efficient bead recovery, ensuring reproducible results
  • High performance membrane -
More Product Information