Pallflex® filters
供应商: CYTIVA PALL
7194 7200 7201 7204 7205 7217 7219 7223 7224 7203 7221 7222 7202
PALL7194EA 0 CNY
PALL7194 PALL7200 PALL7201 PALL7204 PALL7205 PALL7217 PALL7219 PALL7223 PALL7224 PALL7203 PALL7221 PALL7222 PALL7202
Pallflex® filters
过滤器 玻璃纤维过滤器
Versatile filters uniquely suited for a broad range of air monitoring applications.
  • Can be used for high temperature applications
  • Ideal for diesel and other emissions testing, required by environmental regulatory agencies
Tissuquartz™: Pure quartz, no binder. Aerosol retention: 99,9%
Fiberfilm™: Heat resistant borosilicate glass fibres coated with fluorocarbon (TFE). Aerosol retention: 96,4%
Emfab™: Borosilicate glass microfibres reinforced with woven glass cloth and bonded with PTFE. Aerosol retention: 99,95%
* non heat treated