Syringe filters, Acrodisc®, Supor® membrane
供应商: CYTIVA PALL
4187 4504 4506 4508 4509 4525 4602 4604 4611 4612 4614 4618 4650 4651 4652 4653 4654 4655 4656 4658 4659 4661 4662 4608 AP-4426 4668 AP-4425
PALL4187EA 0 CNY
PALL4187 PALL4504 PALL4506 PALL4508 PALL4509 PALL4525 PALL4602 PALL4604 PALL4611 PALL4612 PALL4614 PALL4618 PALL4650 PALL4651 PALL4652 PALL4653 PALL4654 PALL4655 PALL4656 PALL4658 PALL4659 PALL4661 PALL4662 PALL4608 PALLAP-4426 PALL4668 PALLAP-4425
Syringe filters, Acrodisc®, Supor® membrane
过滤器 注射器过滤器
Acrodisc™ syringe filters with Supor™ membrane deliver high flow rates and low protein binding.
  • Superior flow rates and higher throughputs than competitive devices
  • Low protein binding to minimize sample loss
  • Pre-sterilized products are sterilized by gamma irradiation to eliminate potential contamination by EtO residuals
  • A range of sizes (13 to 37 mm) accommodates sample volumes from < 10 to 150 ml
  • Acrodisc PF and Serum Acrodisc syringe filters feature built-in prefilter for increased throughput of difficult-to-filter liquids
  • Reduces mycoplasma with the
More Product Information