Positive pressure filters, AcroCap™, Supor® membrane
供应商: CYTIVA PALL
4480 4482
PALL4480EA 0 CNY
PALL4480 PALL4482
Positive pressure filters, AcroCap™, Supor® membrane
过滤器 瓶口过滤器
Supor® hydrophilic polyethersulfone (PES) membrane, acrylic housing: 0,2 µm vent. Positive pressure filtration of aqueous laboratory solutions. For sterilisation or clarification of up to three litres of serum-free cell and tissue culture media, media additives and other aqueous solutions. Ideal for sterilisation of solutions that tend to foam when filtered under vacuum.
  • Integral hydrophobic vent prevents air lock
  • Reduce mycoplasma using 0,1 µm pore size membrane
  • Sterilised by gamma irradiation to eliminate the risk of cytotoxic residuals