Filter holder, SolVac®
供应商: CYTIVA PALL
4020
PALL4020EA 0 CNY
PALL4020
Filter holder, SolVac®
过滤器支架
Simplifies clean-up and degassing of mobile phase solvents and other solutions. The versatile design fits most HPLC bottles, flasks, and containers, and eliminates the added steps of washing flasks and transferring mobile phase solvent from flask to reservoir.
  • Draws directly from HPLC solvent bottle. Less likely to spill aggressive solvents than glass funnels or disposable cups
  • Patented magnetic seal is reliable and leak proof. Eliminates the possibility of membrane shifting or tearing which can occur with aluminium clamps or threaded holders
现在订购
规格
过滤面积 10,2 cm²
Environmentally Preferable
入口/出口连接 Tapered inlet accepts 3,2 - 6,4 mm ID tubing; outlet seals to bottles with openings 17,8 mm ID to 48,3 mm OD
最高运行温度 38 °C