Carbon Capsule Filter

供应商: CYTIVA PALL
12011
PALL12011EA 0 CNY
PALL12011
Carbon Capsule Filter
过滤器 排气过滤器
Removes odor, colloids, and organics from liquids using activated carbon.

  • Finely divided carbon provides an active surface area of 65000 m²
  • Ready to use and disposable

Contains 100 g of acid-rinsed, activated charcoal. Inlet/outlet connections are ³/₈" FNPT. Includes optional hose barb adapters to accept ¹/₂" int.Ø tubing.

Applications:
- For laboratory and reagent-grade water and chemical filtration
- Protect laboratory from organic solvent odor
现在订购

规格

Environmentally Preferable
最大压力 3,4 bar
最高温度 80 °C
入口/出口 3/8" FNPT; includes optional hose barb fittings to accept 12,7 mm ID tubing

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR