Latex tests, Dryspot
供应商: THERMO FISHER DIAGNOSTICS
DR1108A
OXOIDR1108AEA 0 CNY
OXOIDR1108A
Latex tests, Dryspot
微生物学测试系统 抗原抗体测试 Ag-AK, Latex
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
  • Simplified room temperature storage allows storage for up to two years
  • Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards