Accessories for Anaerobic Systems, Oxoid
供应商: THERMO FISHER DIAGNOSTICS
AN0025A BR0055B
OXOIAN0025AEA 0 CNY
OXOIAN0025A OXOIBR0055B
Accessories for Anaerobic Systems, Oxoid
厌氧系统
Where an AnaeroGen sachet is placed in a sealed jar, the atmospheric oxygen in the jar is rapidly absorbed with the simultaneous generation of carbon dioxide. This novel method differs from others commonly used in that the reaction proceeds with no evolution of hydrogen, and therefore, does not require a catalyst. Furthermore, water is not required to activate the reaction. When used as directed, the AnaeroGen sachet will reduce the oxygen level in the jar to below 1% within 30 minutes. The resulting carbon dioxide level will be between 9% and 13%.
CampyGen Compact for 1 or 2 Petri dishes, isMore Product Information