Large horizontal gel electrophoresis systems, Owl™ A2 and A5
供应商: THERMO OWL SCIENTIFIC
A2 A2-BP A5
OWLAA2EA 0 CNY
OWLAA2 OWLAA2-BP OWLAA5
Large horizontal gel electrophoresis systems, Owl™ A2 and A5
电泳系统 水平凝胶电泳系统
The Owl™ A2 horizontal gel system is a simple, convenient and fast system for detailed DNA/RNA analysis on multiple samples. It is available with or without buffer exchange ports. The Owl A5 large gel system with built-in recirculation prevents formation of pH and ionic gradients for high resolution and uniform reproducible results. The Owl A5 is ideal for long runs, multiple sample sets or RNA gels. It also eliminates uneven migration, band distortion or disassociation of pH dependent glycoxylated RNA molecules that can result when ionic depletion occurs.
  • Each side of the UV-
More Product Information