pH/mV/ORP/°C meters, bench top, Orion Star™ A211
供应商: THERMO ORION
STARA2110
ORIOSTARA2110EA 0 CNY
ORIOSTARA2110
pH/mV/ORP/°C meters, bench top, Orion Star™ A211
电化学仪器 多参数测量仪
Easy to use and budget friendly instrument for a wide range of applications and advanced pH analysis in the lab. Ideal for simple, routine pH, mV and temperature measurements.
  • Performs up to a 5-point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Eliminate the need for magnetic stirring bars using the meter-controlled stirrer probe to uniformly mix solutions
  • Data log collects and