pH buffer solutions, NIST traceable, Orion™

供应商: THERMO ORION

910112 910168 910104 910107 910110
ORIO910112EA 0 CNY
ORIO910112 ORIO910168 ORIO910104 ORIO910107 ORIO910110
pH buffer solutions, NIST traceable, Orion™
缓冲(液) 酸碱缓冲液

Perform accurate, reproducible pH calibrations with Orion™ pH buffers.


  • NIST traceability
  • Colour-coded solutions are available for ease of use and identification


Solutions are manufactured under ISO 9000 quality standards.


The kit includes 1 pint (475 ml) bottle each of pH 4,00 buffer, pH 7,00 buffer, pH 10,01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.


Delivery information: Supplied in 475 ml or 60 ml bottles, with buffer/temperature tables to ensure more precise calibration.

现在订购

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR