pH buffer solutions, NIST traceable, Orion™
供应商: THERMO ORION
910112
910168
910104
910107
910110
ORIO910112EA
0
CNY
ORIO910112
ORIO910168
ORIO910104
ORIO910107
ORIO910110
pH buffer solutions, NIST traceable, Orion™
缓冲(液)
酸碱缓冲液
Perform accurate, reproducible pH calibrations with Orion™ pH buffers.
- NIST traceability
- Colour-coded solutions are available for ease of use and identification
Solutions are manufactured under ISO 9000 quality standards.
The kit includes 1 pint (475 ml) bottle each of pH 4,00 buffer, pH 7,00 buffer, pH 10,01 buffer and standard pH electrode storage solution and one pH electrode storage bottle.
Delivery information: Supplied in 475 ml or 60 ml bottles, with buffer/temperature tables to ensure more precise calibration.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...