Inverted microscope, CKX53
供应商: OLYMPUS OPTICAL
CKX53TR-PH
OLYMCKX53TR-PHEA 0 CNY
OLYMCKX53TR-PH
Inverted microscope, CKX53
显微镜
Ergonomically designed and enhanced imaging for cell culture, the CKX53 is designed for cell sampling and manipulation, especially for fluorescence observations.
  • Observations in lightfield transmission and phase contrast evolutionary to fluorescence
  • High contrast thanks to integrated phase system (IPC) (without ring change)
  • Three-position fluorescence illuminator: Two sets of filters for excitation (blue and green), empty position, manual shutter and neutral filter slide 6%
Compatible with CKX3-SLP and IX2-SL condenser slides, CKX3-SPLIC, CKX3-SLPAS,More Product Information