Flasks on slides, Nunc™
供应商: Thermo Fisher Scientific
170920 177453
NUNC170920EA 0 CNY
NUNC170920 NUNC177453
Flasks on slides, Nunc™
载玻片和盖玻片 载玻片
Glass (Flaskette), PS (SlideFlask), sterile for cell culture on a microscope slide.
  • PS SlideFlask Nunclon™Δ certified
  • SlideFlask is ultrasonically welded to the slide, and individually leak tested
For karyotyping of cells, single cell autoradiography and single cell immuno-fluorescence.
W×D (Flaskette): 20×52 mm; W×D (SlideFlask): 18×50 mm