Chamber slides, Nunc™ Lab-Tek™ II CC2
供应商: Thermo Fisher Scientific
154852 154917 154941 154739
NUNC154852EA 0 CNY
NUNC154852 NUNC154917 NUNC154941 NUNC154739
Chamber slides, Nunc™ Lab-Tek™ II CC2
载玻片和盖玻片 载玻片
Borosilicate coverglass. PS chamber. Sterile.
  • Growth surface remains stable without refrigeration
  • Light blue frosted writing area for clear identification
  • Slide separator to lift the medium chamber from the slide in each pack
Chemically coated growth surface on the glass slide, which mimics poly-lysine, providing binding sites for fastidious cells such as neurons.