Cell culture chambers, Nunc™ EasyFill™ cell factories
供应商: Thermo Fisher Scientific
140000 140250 140360 140400 140410
NUNC140000EA 0 CNY
NUNC140000 NUNC140250 NUNC140360 NUNC140400 NUNC140410
Cell culture chambers, Nunc™ EasyFill™ cell factories
生物反应器
PS, sterile. The EasyFill™ Cell Factory has one large and one small opening in each unit, which makes it versatile and easy to use. EasyFill™ Cell Factory bridges the gap between small scale research and large scale GMP production. It can be used as it is, without any accessories, or used with plug and play connections for rapid attachment of tubing and filters, significantly reducing contamination risk. EasyFill™ systems are available with 1, 2, 4 or 10 growth chamber levels, providing surface areas ranging from 630 to 6300 cm².
  • Versatile - large opening facilitates the desire to pour
More Product Information