Anti-TLR2 Mouse Monoclonal Antibody [clone: TL2.1]

供应商: NOVUS BIOLOGICALS
NB100-56722
NOVUNB100-56722EA 0 CNY
NOVUNB100-56722
Anti-TLR2 Mouse Monoclonal Antibody [clone: TL2.1]
抗体
Type: Primary
Antigen: TLR2 (toll-like receptor 2)
Clonality: monoclonal
Clone: TL2.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
现在订购

规格

抗原 Toll-like Receptor 2,TLR2
Environmentally Preferable
抗原 Toll-like Receptor 2,TLR2
抗原同义词 CD282|TIL4
抗体类型 Primary
克隆形成能力 Monoclonal
结合 Unconjugated
克隆 TL2.1
反应性
主机 Mouse
基因 7097
同位型 IgG2a
免疫沉淀反应法 Yes
荧光免疫检验法 Yes
流式细胞计 Yes
存储温度 Cool
装运温度 Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR