PIG® universal mats
供应商: NEW PIG
MAT203 MAT240 MAT251
NEWPMAT203EA 0 CNY
NEWPMAT203 NEWPMAT240 NEWPMAT251
PIG® universal mats
泄露控制装置
These mats last 2 times longer than ordinary mats so it requires fewer change-outs. Save money with the mat that absorbs anything and works anywhere.
  • Eight layers of 100% PP are thermally bonded to make PIG Mat exceptionally strong; won't rip, tear or fray even when saturated
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave liquids or fibre residue behind
  • Absorbs and retains oils, coolants, solvents and water
  • Dark grey colour hides grime so mat stays on the job
More Product Information