Jars, multipurpose, with covers, Nalgene®
供应商: Thermo Fisher Scientific
DS5300-0507 DS5300-9609 DS5300-9910
NALGDS5300-0507EA 0 CNY
NALGDS5300-0507 NALGDS5300-9609 NALGDS5300-9910
Jars, multipurpose, with covers, Nalgene®
通用罐
Polycarbonate, clear.
  • Drill, tap or apply clamps to jars to attach heating or cooling equipment
  • Very durable
  • Autoclavable
Multipurpose jars with cover, which can be used as water baths, aquaria/terraria or freeze-drying chambers.