Liquid detergent, L900, Nalgene®
供应商: Thermo Fisher Scientific
900-4000
NALG900-4000EA 0 CNY
NALG900-4000
Liquid detergent, L900, Nalgene®
清洗剂 洗涤剂和表面清洁剂
Specifically formulated to clean plastic and glass labware used in pharmaceutical, medical research, academic, biotechnology and other facilities.
  • Formulated to be compatible with all plastics, especially polycarbonate
  • Will not cause crazing, stress cracking or clouding when used as directed
  • For hand or machine use, it is non toxic and low foaming
Non inhibitory according to APHA Standard Methods for the Examination of Water and Wastewater.
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 NAR Certificate of Retirement