Sterilising pans, Nalgene®
供应商: Thermo Fisher Scientific
6902-1000 6902-2000 6902-3000 6902-5000
NALG6902-1000EA 0 CNY
NALG6902-1000 NALG6902-2000 NALG6902-3000 NALG6902-5000
Sterilising pans, Nalgene®
盘子/皿/盆 消毒盘/锅
PP.
  • Heavy duty construction
  • Extra strong rims
For repeated autoclaving of instruments and labware.