Biohazard waste bins, Nalgene®
供应商: Thermo Fisher Scientific
6370-0004 6370-0005 6370-0015
NALG6370-0004EA 0 CNY
NALG6370-0004 NALG6370-0005 NALG6370-0015
Biohazard waste bins, Nalgene®
箱柜 垃圾箱
PP.
  • Port opening in cover for convenient one handed disposal of materials. Less exposure to infectious waste
  • One step sterilisation: No need to remove autoclavable bag
  • Leakproof side and bottom construction
Autoclavable waste containers for secondary containment of biohazardous waste materials such as needles. Holds standard size autoclavable bags. Will not rust, dent or puncture. Red universal biohazard symbol.
Comply with US OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
Certifications: Comply with USMore Product Information