Nalgene™ Staining Boxes
供应商: Thermo Fisher Scientific
5705-1010
NALG5705-1010EA 0 CNY
NALG5705-1010
Nalgene™ Staining Boxes
染色箱
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods.
  • Polymethylpentene box and polypropylene copolymer cover
  • Useful over a –70 to +70 °C (–94 to +158 °F) temperature range
  • Good chemical compatibility with acids, bases and select organic solvents for a versatile range of protocols
  • LDPE plug in bottom for easy drainage of liquid with minimal handling of gel and provides a leakproof seal
  • Plug won't interfere with cover or contents of boxes on a benchtop or shaker
  • Stackable, with a handle on
More Product Information
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 IRECS; ISO 14001 (Certificate Registration No. EMS 752603)