Jars with cover, Nalgene®
供应商: Thermo Fisher Scientific
5352-0002 5352-0004
NALG5352-0002EA 0 CNY
NALG5352-0002 NALG5352-0004
Jars with cover, Nalgene®
通用罐
PP, translucent.
  • Straight sides and no neck making access and retrieval easy
  • Larger sizes make excellent waste crocks
  • Autoclavable
Ideal for storage and transport of solids.