Büchner funnels, Nalgene®
供应商: Thermo Fisher Scientific
4280-0550 4280-0700 4280-0900 4280-1100 4280-1500
NALG4280-0550EA 0 CNY
NALG4280-0550 NALG4280-0700 NALG4280-0900 NALG4280-1100 NALG4280-1500
Büchner funnels, Nalgene®
漏斗 过滤漏斗 Büchner Funnels
PP
  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning
  • Autoclavable
For filter paper or membrane filtration discs.
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 IRECS; ISO 14001 (Certificate Registration No. EMS 752603)