Media bottles, square, sterile, with screw cap, Nalgene®
供应商: Thermo Fisher Scientific
382019-0030 382019-0125 382019-0060 382019-0250 382019-0500 382019-1000 382019-2000
NALG382019-0030EA 0 CNY
NALG382019-0030 NALG382019-0125 NALG382019-0060 NALG382019-0250 NALG382019-0500 NALG382019-1000 NALG382019-2000
Media bottles, square, sterile, with screw cap, Nalgene®
培养基瓶
Certified clean container is made of PETG and comes with a sterile, natural HDPE closure with a tamper-resistant seal.
  • Non-cytotoxic, non-pyrogenic, non-hemolytic; gamma irradiated to a 10-⁶ Sterility Assurance Level
  • Leakproof bottle is free of animal derived components (ADCF) and lot certified for low particulate limits
  • Sterile
Manufactured in a certified ISO 14644-1 Class 7 cleanroom and packaged in cleanroom grade heat-sealed bags to assure product integrity. Suitable for pharmaceutical and biotechnology applications that require processing andMore Product Information