Nalgene® Diagnostics Bottles, Narrow Neck, Round, HDPE, Brown, without Screw Caps
供应商: Thermo Fisher Scientific
362004-9125 362004-9050 362004-9025
NALG362004-9125EA 0 CNY
NALG362004-9125 NALG362004-9050 NALG362004-9025
Nalgene® Diagnostics Bottles, Narrow Neck, Round, HDPE, Brown, without Screw Caps
塑料瓶
Safely store and ship light-sensitive, diagnostic samples and valuable reagents in small volumes with Thermo Scientific™ Nalgene™ opaque amber HDPE diagnostic bottles, used in test kits and many small-volume applications. These opaque, amber, high-density polyethylene bottles are for use with light-sensitive reagents and are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
  • Durable, non-sterile and leakproof
  • Non-pyrogenic and non-cytotoxic
  • Uniform walls are resistant to splitting or puncturing
现在订购
规格
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
环境认证 NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603) NAR Certificate of Retirement