Nalgene® Media Bottles, Square, with Screw Cap
供应商: Thermo Fisher Scientific
322020-0125 322020-0030 322020-0060 322020-0250 322020-0500 322020-1000 322020-2000
NALG322020-0125EA 0 CNY
NALG322020-0125 NALG322020-0030 NALG322020-0060 NALG322020-0250 NALG322020-0500 NALG322020-1000 NALG322020-2000
Nalgene® Media Bottles, Square, with Screw Cap
培养基瓶
PETG bottle with HDPE screw closure.
  • Graduated
  • Durable
  • Break-resistant
  • Leakproof
Package and ship liquid media, buffers and sera using square media bottles with screw caps. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera. Non-sterile bottle offers excellent gas barrier properties; clear PETG material allows for easy identification of contents.
Certifications: USP Class VI, FDA Compliance 21 CFR
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 NAR Certificate of Retirement