Bottles, wide neck, square, with screw cap, Nalgene®
供应商: Thermo Fisher Scientific
2122-0010 2123-0010
NALG2122-0010EA 0 CNY
NALG2122-0010 NALG2123-0010
Bottles, wide neck, square, with screw cap, Nalgene®
塑料瓶
HDPE, translucent with PP screw cap or PPCO, translucent with PP screw cap.
  • HDPE: Compatibility with most alcohols, bases and acids
  • PPCO: Very good chemical resistance, heat resistant up to 121 °C, autoclavable
  • Extra wide opening
Large volume multipurpose bottles for a wide range of applications. Space saving, with convenient integral handles.
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 NAR Certificate of Retirement