Nalgene™ Bottles, Wide Neck, with Screw Cap
供应商: Thermo Fisher Scientific
2120-0005 2120-0010 2121-0005 2120-0010CS
NALG2120-0005EA 0 CNY
NALG2120-0005 NALG2120-0010 NALG2121-0005 NALG2120-0010CS
Nalgene™ Bottles, Wide Neck, with Screw Cap
塑料瓶
HDPE, translucent with PP screw cap or PPCO, translucent with PP screw cap.
  • HDPE: Good chemical resistance, heat resistant up to 120 °C
  • PPCO: Very good chemical resistance, heat resistant up to 121 °C, autoclavable
  • Extra wide opening
Large volume multipurpose bottles for a wide range of uses. Not leakproof.
These high-density, polyethylene bottles are good for use in the freezer to −100 °C (−148 °F). Chemical-resistant HDPE is compatible with a wide variety of lab reagents.
Linerless cap closes the bottle without a liner that can wrinkle, causeMore Product Information
现在订购
规格
Environmentally Preferable Low Manufacturing Impact
环境认证 NAR Certificate of Retirement