Muffle furnaces with lift up door, Tmax 1300 °C and controller C550, LT 5/13 - LT 15/13
供应商: NABERTHERM
L-055H3ANO L-055H3ANOG L-055H3ANOGT L-055H3ANOGTZ L-055H3ANOGZ L-055H3ANOT L-055H3ANOTZ L-055H3ANOZ
NABEL-055H3ANOEA 0 CNY
NABEL-055H3ANO NABEL-055H3ANOG NABEL-055H3ANOGT NABEL-055H3ANOGTZ NABEL-055H3ANOGZ NABEL-055H3ANOT NABEL-055H3ANOTZ NABEL-055H3ANOZ
Muffle furnaces with lift up door, Tmax 1300 °C and controller C550, LT 5/13 - LT 15/13
干燥箱
These furnaces feature heating elements on supporting tubes that radiate freely into the chamber, ensuring particularly short heating times. With the robust lightweight refractory brick insulation, they can reach a max. working temperature of 1300 °C.
  • Heating from two sides
  • Multi-layer insulation with robust lightweight refractory bricks in the furnace chamber
  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Lift up door (LT) with hot surface facing away from the operator
  • Exhaust air outlet on rear wall
More Product Information