T-Mop™ Cleanroom Mop Covers
供应商: MICRONOVA
QDT-18 T6-14 T6-18 QDST-14 T6-18IR QDT-14 TAA-18 TAA-14 T612-18 T612-14 TAA12-14 T612-14IR T20-14 TMC6-14 QPT-18 QPT-14
MICRQDT-18EA 0 CNY
MICRQDT-18 MICRT6-14 MICRT6-18 MICRQDST-14 MICRT6-18IR MICRQDT-14 MICRTAA-18 MICRTAA-14 MICRT612-18 MICRT612-14 MICRTAA12-14 MICRT612-14IR MICRT20-14 MICRTMC6-14 MICRQPT-18 MICRQPT-14
T-Mop™ Cleanroom Mop Covers
拖把
A series of rounded T-mop covers with snaps that easily roll down walls, as well as clean awkward corners, tight spaces, and under equipment. Available in a range of fabrics for specialised cleaning.
  • Low profile, rounded covered sponges
  • Excellent on all room surfaces and equipment exteriors
  • Easily fits in awkward corners, tight spaces, and under equipment
  • Holds up to strong disinfectants
  • Autoclavable
Selection of fabrics include:
NovaPoly™ - 100% smooth knit polyester provides excellent disinfectant dispersal
PolySorb™ - 100% textured polyester forMore Product Information