Accessories for mops micromop™ LoopMop™ and PolyGen™
供应商: MICRONOVA
QDMH-1
MICRQDMH-1EA 0 CNY
MICRQDMH-1
Accessories for mops micromop™ LoopMop™ and PolyGen™
拖把
Universal electropolished stainless steel handles with a quick disconnect connection that works with the entire range of Micronova adapters, tools and mop heads for simplified and versatile cleaning.
  • Universal handles work with all Quick Disconnect tools and mop adapters
  • 304 electropolished stainless steel with polyketone components
  • Strong, hard-wearing connections for long lasting use
  • Available in a range of fixed or extendable lengths from 8 in (20 cm) to 10 ft (3 m)
  • Quick Disconnect end attaches directly to QD mops, tools and adapters
  • SSA-3 adapter
More Product Information