Cleanroom tapes, PCX™
供应商: MICRONOVA
PCX-2DB PCX-2YE PCX-2OR PCX-1BL PCX-2WH PCX-1WH
MICRPCX-2DBEA 0 CNY
MICRPCX-2DB MICRPCX-2YE MICRPCX-2OR MICRPCX-1BL MICRPCX-2WH MICRPCX-1WH
Cleanroom tapes, PCX™
胶带
These cleanroom tapes are made of LDPE with acrylic adhesive.
  • Labeling
  • Sealing, wafer box sealing
  • Over labeling (protect paper labels in water baths)
A low density polyethylene tape, low in halogens and leachable chlorides. Processed and packaged in a cleanroom. The polyethylene film and acrylic adhesive lessen the chance of outgassing. PCX offers excellent chemical resistance and good conformability. Works well with polycarbonates, glass and acrylics. Suitable for the following:
Length: 32,9 m
Different colours available.