MZone™ FlatMop™ Slip-On Mop Covers
供应商: MICRONOVA
MZSM11-16S MZSM11-16SIR MZSM11-9SIR MZSM56-14SIR MZSM56-18S MZSM56-18SIR MZSM620-14SIR
MICRMZSM11-16SEA 0 CNY
MICRMZSM11-16S MICRMZSM11-16SIR MICRMZSM11-9SIR MICRMZSM56-14SIR MICRMZSM56-18S MICRMZSM56-18SIR MICRMZSM620-14SIR
MZone™ FlatMop™ Slip-On Mop Covers
拖把
These slip-on, tab removal flat head mop covers are ideal for cleaning and sanitizing all surfaces and equipment in the cleanroom. A choice of fabric options provide excellent absorbency, effective cleaning, and disinfectant dispersal for specialized cleaning.
  • Easy to change covers fit securely over mop frame
  • Urethane foam interiors fully encased in a choice of fabric:
  • NovaPoly - 100% smooth knit polyester provides excellent disinfectant dispersal
  • PolySorb - 100% textured polyester for removing residues and dispersing disinfectant
  • MegaTex - 100% non-woven
More Product Information