Cleanroom tapes, CR100PC™
供应商: MICRONOVA
CR100PC-12DB CR100PC-2DB CR100PC-2WH CR100PC-1YE CR100PC-1DG_U CR100PC-2DG CR100PC-2RE CR100PC-1TR CR100PC-2TR CR100PC-1WH CR100PC-15DB_U CR100PC-3RE CR100PC-3WH CR100PC-15TR CR100PC-15WH CR100PC-15YE CR100PC-1OR_U CR100PC-15DB CR100PC-1DG CR100PC-1OR CR100PC-1RE CR100-12WHIRM
MICRCR100PC-12DBEA 0 CNY
MICRCR100PC-12DB MICRCR100PC-2DB MICRCR100PC-2WH MICRCR100PC-1YE MICRCR100PC-1DG_U MICRCR100PC-2DG MICRCR100PC-2RE MICRCR100PC-1TR MICRCR100PC-2TR MICRCR100PC-1WH MICRCR100PC-15DB_U MICRCR100PC-3RE MICRCR100PC-3WH MICRCR100PC-15TR MICRCR100PC-15WH MICRCR100PC-15YE MICRCR100PC-1OR_U MICRCR100PC-15DB MICRCR100PC-1DG MICRCR100PC-1OR MICRCR100PC-1RE MICRCR100-12WHIRM
Cleanroom tapes, CR100PC™
胶带
Extremely conformable, vinyl cleanroom tape with synthetic rubber, clean-peel adhesive; ideal for colour coding, sealing and marking on curved and irregular surfaces in cleanroom maintenance and support areas.
  • Clean peel from wafer cartridges and cleanroom work surfaces
  • Synthetic rubber adhesive ensures a clean peel from most cleanroom surfaces such as polyethylene, plastics and stainless steel
  • Matte finish can be written on with a cleanroom pen
  • Ideal for taping and sealing curved and irregular surfaces
  • Special processing and packaging techniques ensure