Bucket Liners
供应商: MICRONOVA
BKL1-1818IR BKL-2430IR BKL-2424IR BKL7-3119 BKL99-3521IR BKL99-3521
MICRBKL1-1818IREA 0 CNY
MICRBKL1-1818IR MICRBKL-2430IR MICRBKL-2424IR MICRBKL7-3119 MICRBKL99-3521IR MICRBKL99-3521
Bucket Liners
废物处理袋
The unique LDPE/nylon laminate is a good barrier material for transporting raw materials or to line buckets containing disinfectants or other cleaning solutions.
  • Laminate is 75% LDPE, 25% nylon
  • Excellent for disinfectants and cleaning solutions
  • Ideal for product parts transport
Designed for cleanrooms and controlled environments. Available as sterile and non-irradiated.
Packaging: Individually double packed, packed in a case of 24/50/200.