Buckets, SlimLine™
供应商: MICRONOVA
B-7DB B-7RE B-7YE B-7DGAC B-7REAC B-7YEAC B-7DG
MICRB-7DBEA 0 CNY
MICRB-7DB MICRB-7RE MICRB-7YE MICRB-7DGAC MICRB-7REAC MICRB-7YEAC MICRB-7DG
Buckets, SlimLine™
桶类
Rectangular, 22 L buckets with a 482,6 mm interior width are an ideal choice for use with a variety of flat head mops and string mops in controlled environments. Available with or without heavy-duty 76,2 mm castors.
  • 6 gallon capacity
  • Volume graduations with gallon and litre marks
  • Colour coded with a choice of red, yellow, blue, green and grey
  • B-7: Use as replacement buckets for SlimLine™ and Slim T™ series carts
  • B-7AC: Use as replacements in BucketBinder™ multi-bucket systems
  • Autoclavable
Micronova Slim T™ polypropylene buckets are designed for stand-More Product Information