Masterflex® Stepping Motor Proportioning Valves, Avantor®
供应商: MASTERFLEX PREMIUM
98651-02 98651-04 98651-08 98651-00 98651-06 98651-10
MFLB98651-02EA 0 CNY
MFLB98651-02 MFLB98651-04 MFLB98651-08 MFLB98651-00 MFLB98651-06 MFLB98651-10
Masterflex® Stepping Motor Proportioning Valves, Avantor®
阀门 Solenoid Valves
Resist pressures up to 500 psi!
  • High precision, two-way metering valves in aluminum or 316 SS for air/water
  • Operate continuously without coil overheating problems found in traditional solenoid designs
Stainless steel models include a 316 SS valve blocks, PTFE-lined aluminum housing blocks, Viton® O-rings, and PFA closing pins.
Aluminum models include aluminum housings and valve blocks, Viton® O-rings, and PFA closing pins.
Also includes a 9-pin "D" electrical connector. Optional connection cable (98651-50) for easy input/output wiring. Power input is +1More Product Information