Warren Rupp Air-Operated Double Diaphragm Pumps
供应商: MASTERFLEX PREMIUM
74440-50
MFLB74440-50EA 0 CNY
MFLB74440-50
Warren Rupp Air-Operated Double Diaphragm Pumps
Air-Operated Pumps
Complies for applications in food processing, pharmaceutical, and cosmetic applications.
  • Handle abrasive and shear-sensitive materials
  • Self-priming up to 24 ft of water
  • Deadhead against closed discharge without damage
These Warren Rupp Air-Operated Double Diaphragm Pumps are available in two compliance models. The USDA certified pump is ideal for clean-in-place sanitary installations. The FDA-compliant pumps are ideal for food processing, pharmaceutical, and cosmetic applications.