Tygon® E-1000 Ultra-Soft Flexible Transfer Tubing

供应商: MASTERFLEX PREMIUM
65102-00 65102-02 65102-04 65102-06 65102-08 65102-16 65102-10 65102-14
MFLB65102-00EA 0 CNY
MFLB65102-00 MFLB65102-02 MFLB65102-04 MFLB65102-06 MFLB65102-08 MFLB65102-16 MFLB65102-10 MFLB65102-14
Tygon® E-1000 Ultra-Soft Flexible Transfer Tubing
胶管 Transfer Tubing
High degree of flexibility, performs well at low temperatures.

  • Global market leader in range of formulations for most extensive chemical compatibility

Formulation: Tygon® E-1000
Applications: General laboratory, vent and drain lines. Ideal for lab installations requiring multiple directional changes, abrupt curves, or twisting.
Characteristics: High degree of flexibility, performs well at low temperatures, broad chemical resistance, resists kinking and collapse. Transparent.
Certification: FDA compliant. Made according to GMP.
Temperature range: –67 to 125More Product Information
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR